Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043317.2 | NP_001036782.1 | 16 | 2850 | ||
NM_009845.3 | NP_033975.3 | 14 | 2725 | |||
GenBank mRNA | AF102134.1 | - | 13 | 2494 | ||
AF115400.1 | - | 13 | 2494 | |||
AF115401.1 | - | 13 | 2494 | |||
AK041230.1 | - | 16 | 2857 | |||
AK143835.1 | - | 16 | 2788 | |||
AK156735.1 | - | 14 | 2724 | |||
AK171589.1 | - | 13 | 2531 | |||
AK172301.1 | - | 14 | 2681 | |||
BC046957.1 | - | 16 | 2795 | |||
BC150807.1 | - | 15 | 2809 | 9087718 | ||
L02844.1 | - | 15 | 2996 | |||
L16928.1 | - | 15 | 2973 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK145736.1 | - | 1099 | Chr. - - [-] on Build |