Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159555.1 | NP_001153027.1 | 6 | 964 | ||
NM_001159556.1 | NP_001153028.1 | 6 | 824 | |||
NM_001159557.1 | NP_001153029.1 | 7 | 875 | |||
NM_001159558.1 | NP_001153030.1 | 7 | 991 | |||
NM_007643.4 | NP_031669.3 | 6 | 967 | |||
GenBank mRNA | AK052825.1 | - | 6 | 768 | ||
AK128908.1 | - | 7 | 768 | |||
AK150648.1 | - | 6 | 788 | |||
AK150865.1 | - | 2 | 162 | |||
AK151355.1 | - | 6 | 803 | |||
AK152904.1 | - | 4 | 474 | |||
AK153095.1 | - | 1 | 77 | |||
AK172162.1 | - | 7 | 813 | |||
BC010262.1 | - | 6 | 776 | 3481681 | ||
L23108.1 | - | 6 | 785 |