Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159555.1 | NP_001153027.1 | 10 | 1426 | ||
NM_001159556.1 | NP_001153028.1 | 10 | 1286 | |||
NM_001159557.1 | NP_001153029.1 | 11 | 1337 | |||
NM_001159558.1 | NP_001153030.1 | 11 | 1453 | |||
NM_007643.4 | NP_031669.3 | 10 | 1429 | |||
GenBank mRNA | AK052825.1 | - | 10 | 1230 | ||
AK128908.1 | - | 11 | 1230 | |||
AK150648.1 | - | 10 | 1250 | |||
AK150865.1 | - | 6 | 624 | |||
AK151355.1 | - | 10 | 1265 | |||
AK152642.1 | - | 5 | 421 | |||
AK152904.1 | - | 8 | 936 | |||
AK153095.1 | - | 5 | 539 | |||
AK172162.1 | - | 11 | 1275 | |||
BC010262.1 | - | 10 | 1238 | 3481681 | ||
L23108.1 | - | 10 | 1247 |