Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110320.1 | NP_001103790.1 | 6 | 801 | ||
NM_001110321.1 | NP_001103791.1 | 5 | 729 | |||
NM_001110322.1 | NP_001103792.1 | 4 | 639 | |||
NM_007654.2 | NP_031680.2 | 6 | 801 | |||
GenBank mRNA | AF543213.1 | - | 6 | 702 | ||
AF543214.1 | - | 4 | 477 | |||
AK134132.1 | - | 6 | 804 | |||
AK144191.1 | - | 5 | 732 | |||
AK151094.1 | - | 641 | ||||
BC003824.1 | - | 6 | 759 | 3599554 | ||
J04170.1 | - | 6 | 738 | |||
S40777.1 | - | 6 | 734 |