Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009858.2 | NP_033988.1 | 3 | 484 | ||
GenBank mRNA | AK087944.1 | - | 3 | 461 | ||
AK088372.1 | - | 3 | 509 | |||
AK088649.1 | - | 3 | 508 | |||
AK154055.1 | - | 3 | 464 | |||
AK162993.1 | - | 3 | 473 | |||
AK169581.1 | - | 3 | 485 | |||
M16799.1 | - | 3 | 477 | |||
M17534.1 | - | 3 | 484 | |||
M19504.1 | - | 3 | 472 | |||
X07698.1 | - | 3 | 488 |