Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc25b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111075.4 | NP_001104545.1 | 7 | 948 | ||
NM_023117.4 | NP_075606.1 | 8 | 1026 | |||
GenBank mRNA | AK004836.1 | - | 7 | 905 | ||
AK043730.1 | - | 7 | 939 | |||
AK153903.1 | - | 8 | 1027 | |||
AK157397.1 | - | 7 | 906 | |||
BC048164.1 | - | 7 | 945 | |||
BC057568.1 | - | 7 | 819 | 6401489 | ||
S93521.1 | - | 8 | 895 | |||
W54021.1 | - | 3 | 168 |