Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007661.3 | NP_031687.2 | 11, 12 | 1352 | ||
GenBank mRNA | AK077668.1 | - | 11, 12 | 1353 | ||
AK147133.1 | - | 11, 12 | 1330 | |||
BC052920.1 | - | 11, 12 | 1430 | 6489832 | ||
L37092.1 | - | 10, 11 | 1924 |