Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271566.1 | NP_001258495.1 | 9 | 1271 | ||
NM_001271567.1 | NP_001258496.1 | 8 | 1006 | |||
NM_001271568.1 | NP_001258497.1 | 8 | 1006 | |||
NM_009863.3 | NP_033993.2 | 9 | 1102 | |||
GenBank mRNA | AB018574.1 | - | 8 | 1007 | ||
AB018575.1 | - | 9 | 1103 | |||
AB018576.1 | - | 7 | 1708 | |||
AB018577.1 | - | 9 | 1073 | |||
AJ007661.1 | - | 9 | 1054 | |||
AK145921.1 | - | 9 | 1028 | |||
AK146009.1 | - | 9 | 1273 | |||
BC062149.1 | - | 8 | 1820 | |||
BC072666.1 | - | 9 | 1043 | 30611704 |