Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009867.2 | NP_033997.1 | 10 | 1539 | ||
GenBank mRNA | D14888.1 | - | 10 | 1449 | ||
X69966.1 | - | 10 | 1395 |