Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009868.4 | NP_033998.2 | 11 | 1744 | ||
GenBank mRNA | AK164474.1 | - | 11 | 1727 | ||
AK167620.1 | - | 11 | 1676 | |||
BC054790.1 | - | 11 | 1670 | 6831962 | ||
D63942.1 | - | 11 | 1705 | |||
X83930.2 | - | 11 | 1659 |