Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016756.4 | NP_058036.1 | 1, 2 | 342 | ||
NM_183417.3 | NP_904326.1 | 1, 2 | 342 | |||
GenBank mRNA | AJ223732.1 | - | 1, 2 | 244 | ||
AK146845.1 | - | 1, 2 | 322 | |||
AK148017.1 | - | 1, 2 | 281 | |||
AK151979.1 | - | 1, 2 | 292 | |||
AK152922.1 | - | 1, 2 | 295 | |||
AK153983.1 | - | 1, 2 | 294 | |||
AK154997.1 | - | 1, 2 | 294 | |||
AK179714.1 | - | 13 | ||||
AK181235.1 | - | 13 | ||||
AK184593.1 | - | 13 | ||||
AK186022.1 | - | 13 | ||||
AK186779.1 | - | 13 | ||||
AK187629.1 | - | 1, 2 | 78 | |||
AK189612.1 | - | 1, 2 | 78 | |||
AK210959.1 | - | 13 | ||||
BC005654.1 | - | 1, 2 | 303 | 3709730 | ||
CN705407.1 | - | 1, 2 | 342 | |||
U63337.1 | - | 1, 2 | 270 |