Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009886.2 | NP_034016.2 | 1 | 3456 | ||
GenBank mRNA | AF031572.1 | - | 1 | 3456 |
Set Membership: |
G-protein coupled receptor |