Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009891.2 | NP_034021.1 | 5 | 609 | ||
GenBank mRNA | AK046406.1 | - | 5 | 610 | ||
AK047708.1 | - | 4 | 501 | |||
AK143147.1 | - | 4 | 507 | |||
BC119322.1 | - | 4 | 371 | 8734071 | ||
BC120589.1 | - | 4 | 371 | 40129512 | ||
D12487.1 | - | 4 | 533 | |||
D12490.1 | - | 4 | 502 |