Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007695.3 | NP_031721.2 | 5 | 469 | ||
GenBank mRNA | AK051475.1 | - | 5 | 410 | ||
AK155412.1 | - | 5 | 409 | |||
BC003780.1 | - | 5 | 405 | 3601796 | ||
BC004734.1 | - | 5 | 396 | 3583267 | ||
BC005611.1 | - | 5 | 416 | 3582304 | ||
CT010283.1 | - | 5 | 345 | |||
EU313768.1 | - | 5 | 345 | |||
X93035.1 | - | 5 | 410 |