Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007697.2 | NP_031723.2 | 23, 24 | 3474 | ||
GenBank mRNA | AK048639.1 | - | 7, 8 | 1060 | ||
BC060216.1 | - | 23, 24 | 3440 | |||
BC131670.1 | - | 23, 24 | 3282 | 40110867 | ||
BC131671.1 | - | 23, 24 | 3278 | 40110873 | ||
X94310.1 | - | 23, 24 | 3348 |