Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007697.2 | NP_031723.2 | 27 | 3902 | ||
GenBank mRNA | AK048639.1 | - | 11 | 1488 | ||
AK132840.1 | - | 13 | 1807 | |||
BC060216.1 | - | 27 | 3868 | |||
BC131670.1 | - | 27 | 3710 | 40110867 | ||
BC131671.1 | - | 27 | 3706 | 40110873 | ||
X94310.1 | - | 27 | 3776 |