Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021350.2 | NP_067325.2 | 1 | 274 | ||
GenBank mRNA | BC118932.2 | - | 1 | 274 | 40090670 |