Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007699.2 | NP_031725.1 | 2 | 729 | ||
GenBank mRNA | BC120504.1 | - | 1 | 573 | 40129427 | |
BC137677.1 | - | 1 | 573 | 8860696 |
Set Membership: |
G-protein coupled receptor |