Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007708.3 | NP_031734.3 | 10 | 2043 | ||
GenBank mRNA | AF070066.1 | - | 2 | 214 | ||
AF086823.1 | - | 10 | 2099 | |||
AF086824.1 | - | 10 | 2099 | |||
AK145037.1 | - | 11 | 1399 | |||
CF744826.1 | - | 2 | 201 |