Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn4-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035804.1 | - | 12 | 2330 | ||
AK050833.1 | - | 2 | 101 | |||
AK088317.1 | - | 9 | 3716 | |||
AK133759.1 | - | 13 | 2264 | |||
AK209939.1 | - | 2 | 112 | |||
AK215840.1 | - | 1 | 70 | |||
BC005553.1 | - | 13 | 2298 | 3586154 | ||
BC110668.1 | - | 13 | 2690 | 6391053 | ||
DQ186662.1 | - | 11 | 2188 | |||
Z49916.1 | - | 13 | 2601 |