Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009906.5 | NP_034036.1 | 8 | 922 | ||
GenBank mRNA | AF111172.1 | - | 8 | 930 | ||
AJ011912.1 | - | 8 | 916 | |||
AK002418.1 | - | 8 | 941 | |||
AK048279.1 | - | 8 | 923 | |||
AK083832.1 | - | 7 | 2187 | |||
AK146759.1 | - | 8 | 916 | |||
AK155021.1 | - | 8 | 941 | |||
AK165669.1 | - | 8 | 947 | |||
AK166648.1 | - | 6 | 715 | |||
AK169924.1 | - | 8 | 929 | |||
AK170781.1 | - | 8 | 918 | |||
BC024820.1 | - | 8 | 924 | 5360558 |