Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009906.5 | NP_034036.1 | 11 | 1297 | ||
GenBank mRNA | AF111172.1 | - | 11 | 1305 | ||
AJ011912.1 | - | 11 | 1291 | |||
AK002418.1 | - | 11 | 1316 | |||
AK048279.1 | - | 11 | 1298 | |||
AK083832.1 | - | 10 | 2562 | |||
AK146759.1 | - | 11 | 1291 | |||
AK155021.1 | - | 11 | 1316 | |||
AK165669.1 | - | 11 | 1322 | |||
AK166648.1 | - | 9 | 1090 | |||
AK169924.1 | - | 11 | 1304 | |||
AK170781.1 | - | 11 | 1293 | |||
BC024820.1 | - | 11 | 1299 | 5360558 |