Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111110.2 | NP_001104580.1 | 14 | 1962 | ||
NM_001284519.1 | NP_001271448.1 | 15 | 2124 | |||
NM_001284520.1 | NP_001271449.1 | 12 | 1479 | |||
NM_007717.5 | NP_031743.3 | 13 | 1617 | |||
GenBank mRNA | AB061276.1 | - | 14 | 1946 | ||
AB061277.1 | - | 15 | 2108 | |||
AK083394.1 | - | 12 | 1495 | |||
AK145110.1 | - | 12 | 1766 | |||
AK166486.1 | - | 13 | 1618 | |||
BC028474.1 | - | 14 | 1972 | |||
BC055079.1 | - | 11 | 1476 | 30077348 | ||
D21826.1 | - | 12 | 1589 |