Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009908.2 | NP_034038.2 | 5 | 813 | ||
GenBank mRNA | AJ006215.1 | - | 5 | 726 | ||
AK087150.1 | - | 2 | 115 | |||
AK219252.1 | - | 2 | 111 | |||
BC031500.1 | - | 5 | 742 | 2647633 | ||
BC063776.1 | - | 5 | 742 | 6398450 |