Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009908.2 | NP_034038.2 | 4 | 667 | ||
GenBank mRNA | AJ006215.1 | - | 4 | 580 | ||
AK155730.1 | - | 4 | 618 | |||
BC031500.1 | - | 4 | 596 | 2647633 | ||
BC063776.1 | - | 4 | 596 | 6398450 |