Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ackr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271607.1 | NP_001258536.1 | 3 | 1275 | ||
NM_007722.4 | NP_031748.2 | 2 | 992 | |||
GenBank mRNA | AF000236.1 | - | 2 | 946 | ||
AK031100.1 | - | 970 | ||||
BC015254.1 | - | 2 | 907 | 4242244 |