Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146323.1 | NP_001139795.1 | 11 | 1714 | ||
NM_080634.4 | NP_542365.3 | 11 | 2110 | |||
GenBank mRNA | AF393747.1 | - | 11 | 2099 | ||
AF393780.1 | - | 11 | 2102 | |||
AK034377.1 | - | 11 | 2081 | |||
AK036663.1 | - | 11 | 2118 | |||
AK037829.1 | - | 9 | 2546 | |||
AK155497.1 | - | 11 | 2077 | |||
AK157953.1 | - | 11 | 2096 | |||
AK166194.1 | - | 10 | 3187 | |||
AK169855.1 | - | 10 | 2182 | |||
AK170581.1 | - | 11 | 1626 | |||
AK171997.1 | - | 11 | 1702 | |||
BC020105.1 | - | 1 | 209 | |||
BC038087.1 | - | 1 | 201 | |||
BC049350.1 | - | 2 | 301 | |||
BC058235.1 | - | 11 | 2084 | 6307400 |