Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109991.1 | NP_001103461.1 | 33 | 3394 | ||
NM_009929.3 | NP_034059.2 | 34 | 2810 | |||
GenBank mRNA | BC043697.1 | - | 33 | 3394 | ||
BC062931.1 | - | 34 | 2777 | |||
BC064817.1 | - | 34 | 2781 | 6332464 | ||
BC066080.1 | - | 34 | 2776 | 30355615 | ||
CF171159.1 | - | 6 | 359 | |||
D17546.1 | - | 32 | 2204 | |||
L16898.1 | - | 34 | 2749 | |||
L22545.1 | - | 32 | 2648 | |||
U03714.1 | - | 4 | 230 |