Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009930.2 | NP_034060.2 | 2 | 322 | ||
GenBank mRNA | AK013329.1 | - | 2 | 230 | ||
AK029212.1 | - | 2 | 226 | |||
AK031544.1 | - | 2 | 195 | |||
AK041115.1 | - | 2 | 230 | |||
AK048546.1 | - | 2 | 230 | |||
AK147542.1 | - | 2 | 228 | |||
AK157762.1 | - | 2 | 230 | |||
AK159763.1 | - | 2 | 230 | |||
AK159920.1 | - | 2 | 230 | |||
AK159985.1 | - | 2 | 230 | |||
AK160048.1 | - | 2 | 230 | |||
AK160107.1 | - | 2 | 229 | |||
AK160818.1 | - | 2 | 230 | |||
AK220558.1 | - | 2 | 245 | |||
BC043089.1 | - | 2 | 197 | 6418299 | ||
BC052398.1 | - | 2 | 214 | 5720872 | ||
BC058724.1 | - | 2 | 231 | 5705762 | ||
BY139268.1 | - | 2 | 322 | |||
M18933.1 | - | 2 | 131 |