Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007734.2 | NP_031760.2 | 50 | 5144 | ||
GenBank mRNA | AF169387.1 | - | 50 | 5089 | ||
AK033387.1 | - | 1 | 51 | |||
X82205.1 | - | 2 | 252 | |||
Z35166.1 | - | 3 | 451 |