Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007735.2 | NP_031761.1 | 8 | 867 | ||
GenBank mRNA | AF169388.1 | - | 8 | 810 | ||
AK142553.1 | - | 8 | 742 | |||
AK164732.1 | - | 8 | 867 | |||
BC117709.1 | - | 7 | 627 |