Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015734.2 | NP_056549.2 | 2 | 579 | ||
GenBank mRNA | AB009993.1 | - | 2 | 319 | ||
BC138447.1 | - | 2 | 253 | 8861467 | ||
BC138449.1 | - | 2 | 253 | 8861469 | ||
BY713878.1 | - | 2 | 581 |