Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243008.1 | NP_001229937.1 | 5 | 1735 | ||
NM_001243009.1 | NP_001229938.1 | 3 | 514 | |||
GenBank mRNA | AF034136.1 | - | 5 | 1692 |