Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243008.1 | NP_001229937.1 | 10 | 4740 | ||
NM_001243009.1 | NP_001229938.1 | 7 | 2919 | |||
GenBank mRNA | AF034136.1 | - | 10 | 4697 | ||
AF064749.1 | - | 7 | 2726 | |||
AK076391.1 | - | 2 | 722 |