Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Comt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111062.1 | NP_001104532.1 | 6 | 1047 | ||
NM_001111063.1 | NP_001104533.1 | 5 | 858 | |||
NM_007744.3 | NP_031770.2 | 5 | 990 | |||
GenBank mRNA | AF076156.1 | - | 5 | 792 | ||
AK148311.1 | - | 6 | 714 | |||
BC010402.1 | - | 5 | 784 | 4210097 |