Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009938.4 | NP_034068.3 | 33 | 4035 | ||
GenBank mRNA | AK032071.1 | - | 11 | 1988 | ||
AK043479.1 | - | 33 | 3761 | |||
AK189379.1 | - | 2 | 135 | |||
AK193412.1 | - | 2 | 135 | |||
AK194620.1 | - | 2 | 145 | |||
AK199106.1 | - | 3 | 339 | |||
AK199738.1 | - | 2 | 135 | |||
BC005609.1 | - | 7 | 883 | |||
BC024070.1 | - | 33 | 3688 | 5097228 | ||
BC025896.1 | - | 15 | 1722 | |||
BC047429.1 | - | 33 | 3726 | 3498656 | ||
BC082785.1 | - | 29 | 3375 |