Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285507.1 | NP_001272436.1 | 12, 13 | 1306 | ||
NM_001285512.1 | NP_001272441.1 | 12, 13 | 1333 | |||
NM_001285513.1 | NP_001272442.1 | 11, 12 | 1097 | |||
NM_009939.3 | NP_034069.2 | 12, 13 | 1285 | |||
GenBank mRNA | AF071312.1 | - | 12, 13 | 1195 | ||
AF087688.1 | - | 12, 13 | 1192 | |||
AK010383.1 | - | 12, 13 | 1232 | |||
AK037757.1 | - | 12, 13 | 1232 | |||
AK084421.1 | - | 12, 13 | 1206 | |||
AK132207.1 | - | 12, 13 | 1202 | |||
AK132776.1 | - | 12, 13 | 1270 | |||
AK145225.1 | - | 11, 12 | 1009 | |||
AK146153.1 | - | 12, 13 | 1196 | |||
BC023096.1 | - | 12, 13 | 1190 | 3675508 |