Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007753.2 | NP_031779.1 | 7 | 631 | ||
GenBank mRNA | AK088906.1 | - | 7 | 632 | ||
BC108409.1 | - | 7 | 620 | |||
J05118.1 | - | 7 | 627 |