Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007757.2 | NP_031783.2 | 4 | 1120 | ||
GenBank mRNA | AK089471.1 | - | 1 | 366 | ||
AK157277.1 | - | 4 | 1003 | |||
BC017680.1 | - | 4 | 977 | 4241687 | ||
CA535096.1 | - | 4 | 448 | |||
D16333.1 | - | 4 | 1034 |