Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013496.3 | NP_038524.1 | 2, 3 | 395 | ||
GenBank mRNA | AK045283.1 | - | 2, 3 | 337 | ||
AK144397.1 | - | 2, 3 | 347 | |||
CD742428.1 | - | 2, 3 | 421 | |||
M31552.1 | - | 1, 2 | 48 | |||
X15481.1 | - | 2, 3 | 321 | |||
X15789.1 | - | 2, 3 | 337 |