Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007760.3 | NP_031786.2 | 5 | 1019 | ||
GenBank mRNA | AK041431.1 | - | 5 | 794 | ||
AK132179.1 | - | 6 | 846 | |||
AK170740.1 | - | 5 | 805 | |||
BC006668.1 | - | 5 | 766 | 3499169 | ||
X85983.1 | - | 5 | 737 |