Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013497.3 | NP_038525.2 | 1 | 210 | ||
GenBank mRNA | AK090077.1 | - | 1 | 180 | ||
AK132785.1 | - | 1 | 220 | |||
AK144208.1 | - | 1 | 213 | |||
BC002094.1 | - | 1 | 176 | 3490360 |