Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crhbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198408.3 | NP_940800.1 | 6 | 922 | ||
GenBank mRNA | AK135067.1 | - | 6 | 923 | ||
AK139404.1 | - | 6 | 768 | |||
BC061247.1 | - | 6 | 863 | 30246702 |