Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007792.4 | NP_031818.3 | 4 | 375 | ||
GenBank mRNA | AF037208.1 | - | 4 | 375 | ||
AK003328.1 | - | 4 | 374 | |||
AK010626.1 | - | 4 | 376 | |||
AK012647.1 | - | 4 | 373 | |||
BC012663.1 | - | 4 | 360 | 4210909 | ||
CK031156.1 | - | 4 | 361 | |||
D88792.1 | - | 3 | 301 |