Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007792.4 | NP_031818.3 | 6 | 618 | ||
GenBank mRNA | AF037208.1 | - | 6 | 618 | ||
AK003328.1 | - | 6 | 617 | |||
AK010626.1 | - | 6 | 619 | |||
AK012647.1 | - | 6 | 616 | |||
BC012663.1 | - | 6 | 603 | 4210909 | ||
CK031156.1 | - | 6 | 604 | |||
D88792.1 | - | 5 | 544 |