Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007798.3 | NP_031824.1 | 5 | 533 | ||
GenBank mRNA | AK002456.1 | - | 5 | 482 | ||
AK083393.1 | - | 5 | 533 | |||
AK147192.1 | - | 5 | 480 | |||
AK149884.1 | - | 5 | 482 | |||
AK151790.1 | - | 5 | 480 | |||
AK153047.1 | - | 5 | 479 | |||
AK159992.1 | - | 5 | 518 | |||
AK167361.1 | - | 5 | 514 | |||
AK170959.1 | - | 5 | 506 | |||
AK179071.1 | - | 1 | 15 | |||
AK184697.1 | - | 1 | 15 | |||
AK192159.1 | - | 1 | 15 | |||
AK192561.1 | - | 1 | 15 | |||
AK197104.1 | - | 1 | 15 | |||
AK198001.1 | - | 1 | 15 | |||
AK198619.1 | - | 1 | 15 | |||
AK198709.1 | - | 1 | 15 | |||
AK201305.1 | - | 1 | 15 | |||
AK201756.1 | - | 1 | 15 | |||
AK202640.1 | - | 1 | 15 | |||
AK203701.1 | - | 1 | 15 | |||
AK205609.1 | - | 1 | 15 | |||
AK213062.1 | - | 1 | 15 | |||
BC006656.1 | - | 5 | 524 | 3500700 | ||
M14222.1 | - | 4 | 438 | |||
S69034.1 | - | 5 | 529 | |||
Y18463.1 | - | 1 | 92 |