Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxadr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025192.3 | NP_001020363.1 | 6 | 935 | ||
NM_009988.4 | NP_034118.1 | 6 | 935 | |||
GenBank mRNA | AK004908.2 | - | 6 | 860 | ||
AK145569.1 | - | 6 | 861 | |||
AK160457.1 | - | 6 | 857 | |||
BC016457.1 | - | 6 | 850 | 4216727 | ||
U90715.1 | - | 6 | 835 | |||
Y10320.1 | - | 6 | 774 | |||
Y11929.1 | - | 6 | 889 |
Gene Symbol: Btg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078652.1 | - | 744 |