Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cybb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007807.5 | NP_031833.3 | 4, 5 | 422 | ||
GenBank mRNA | AK033570.1 | - | 4, 5 | 366 | ||
AK053920.1 | - | 4, 5 | 366 | |||
AK152257.1 | - | 4, 5 | 393 | |||
AK153154.1 | - | 4, 5 | 393 | |||
AK153259.1 | - | 4, 5 | 393 | |||
AK154126.1 | - | 4, 5 | 422 | |||
AK171526.1 | - | 4, 5 | 408 | |||
BC042838.1 | - | 4, 5 | 389 | |||
BC071229.1 | - | 4, 5 | 377 | 4979804 | ||
FJ168469.1 | - | 4, 5 | 411 | |||
U43384.1 | - | 4, 5 | 348 |