Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | K02589.1 | - | 1284 |
Gene Symbol: Cyp1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136059.2 | NP_001129531.1 | 6 | 1343 | ||
NM_009992.4 | NP_034122.1 | 6 | 1422 | |||
GenBank mRNA | AK005000.1 | - | 6 | 1423 | ||
BC125440.1 | - | 6 | 1342 | 40129955 | ||
BC125444.1 | - | 6 | 1342 | 40129959 | ||
K02588.1 | - | 6 | 1342 | |||
Y00071.1 | - | 6 | 1343 |