Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013809.1 | NP_038837.1 | 6 | 916 | ||
GenBank mRNA | AK014160.1 | - | 6 | 934 | ||
BC046321.1 | - | 6 | 951 | |||
BC058222.1 | - | 6 | 921 | 6306453 | ||
L81171.1 | - | 6 | 916 |